Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTTTCTCACTCTTGGCTGCAGG[C/G]CCATCCTGTCTGTGGAGAAGGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GPKOW PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPKOW - G-patch domain and KOW motifs | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015698.5 | 1105 | Silent Mutation | GGC,GGG | G,G 342 | NP_056513.2 | |
XM_017029415.1 | 1105 | Silent Mutation | GGC,GGG | G,G 357 | XP_016884904.1 |