Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCATTGATGCCCGTTTCAAAGA[A/G]ATAAAATATAGAGGGATGTTCCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300242 | ||||||||||||||||||||
Literature Links: |
SLC25A14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC25A14 - solute carrier family 25 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282195.1 | 479 | Silent Mutation | GAA,GAG | E,E 81 | NP_001269124.1 | |
NM_001282196.1 | 479 | Silent Mutation | GAA,GAG | E,E 78 | NP_001269125.1 | |
NM_001282197.1 | 479 | Silent Mutation | GAA,GAG | E,E 78 | NP_001269126.1 | |
NM_001282198.1 | 479 | Silent Mutation | GAA,GAG | E,E 46 | NP_001269127.1 | |
XM_005262489.4 | 479 | Intron | XP_005262546.1 | |||
XM_011531402.2 | 479 | Silent Mutation | GAA,GAG | E,E 90 | XP_011529704.1 | |
XM_011531403.1 | 479 | Intron | XP_011529705.1 | |||
XM_017029936.1 | 479 | Silent Mutation | GAA,GAG | E,E 46 | XP_016885425.1 | |
XM_017029937.1 | 479 | Silent Mutation | GAA,GAG | E,E 90 | XP_016885426.1 | |
XM_017029938.1 | 479 | Silent Mutation | GAA,GAG | E,E 81 | XP_016885427.1 | |
XM_017029939.1 | 479 | Silent Mutation | GAA,GAG | E,E 78 | XP_016885428.1 | |
XM_017029940.1 | 479 | Intron | XP_016885429.1 | |||
XM_017029941.1 | 479 | UTR 5 | XP_016885430.1 | |||
XM_017029942.1 | 479 | Intron | XP_016885431.1 | |||
XM_017029943.1 | 479 | Intron | XP_016885432.1 |