Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATTTTCCATATGCACCTTGGAT[A/G]AATGATGTGGATGTTATTAAAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMGT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMGT1 - membrane magnesium transporter 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9A6 - solute carrier family 9 member A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042537.1 | Intron | NP_001036002.1 | ||||
NM_001177651.1 | Intron | NP_001171122.1 | ||||
NM_006359.2 | Intron | NP_006350.1 | ||||
XM_006724726.3 | Intron | XP_006724789.1 | ||||
XM_011531243.2 | Intron | XP_011529545.1 | ||||
XM_017029223.1 | Intron | XP_016884712.1 | ||||
XM_017029224.1 | Intron | XP_016884713.1 | ||||
XM_017029225.1 | Intron | XP_016884714.1 |