Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATGTCTTCAGCTTTGTGAAGAT[A/T]GCAAAGCTTTCGAGGTAGGGGCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300838 | ||||||||||||||||||||
Literature Links: |
FRMPD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRMPD4 - FERM and PDZ domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014728.3 | 533 | Silent Mutation | ATA,ATT | I,I 9 | NP_055543.2 | |
XM_005274632.3 | 533 | Intron | XP_005274689.2 | |||
XM_011545613.2 | 533 | Silent Mutation | ATA,ATT | I,I 9 | XP_011543915.1 | |
XM_017029983.1 | 533 | Intron | XP_016885472.1 | |||
XM_017029984.1 | 533 | Intron | XP_016885473.1 | |||
XM_017029985.1 | 533 | Intron | XP_016885474.1 | |||
XM_017029986.1 | 533 | Intron | XP_016885475.1 |