Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCTTCTGGAATCAGGAAGCAAG[C/T]AGAATTTTGTCATGGTTTTCTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRDL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRDL1 - chordin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143981.1 | Intron | NP_001137453.1 | ||||
NM_001143982.1 | Intron | NP_001137454.1 | ||||
NM_001143983.2 | Intron | NP_001137455.2 | ||||
NM_145234.3 | Intron | NP_660277.2 | ||||
XM_005262221.1 | Intron | XP_005262278.1 | ||||
XM_005262222.3 | Intron | XP_005262279.1 | ||||
XM_005262223.1 | Intron | XP_005262280.1 | ||||
XM_005262224.1 | Intron | XP_005262281.1 | ||||
XM_017029959.1 | Intron | XP_016885448.1 |