Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGCGACCTGGCCCCTAGCTGCC[C/T]TCTGCGGGGCCGCGACCTCCTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300724 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNKSR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNKSR2 - connector enhancer of kinase suppressor of Ras 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLHL34 - kelch like family member 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153270.2 | 1507 | Missense Mutation | AAG,AGG | K,R 303 | NP_695002.1 |