Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACATGTCAAAGGCCCGGGCCTC[A/G]ATGAGCTCGCCGCTGACCCCTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM3A - family with sequence similarity 3 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171132.2 | 1069 | Silent Mutation | ATC,ATT | I,I 118 | NP_001164603.1 | |
NM_001171133.2 | 1069 | Silent Mutation | ATC,ATT | I,I 80 | NP_001164604.1 | |
NM_001171134.2 | 1069 | Intron | NP_001164605.1 | |||
NM_001282311.1 | 1069 | Silent Mutation | ATC,ATT | I,I 132 | NP_001269240.1 | |
NM_001282312.1 | 1069 | Silent Mutation | ATC,ATT | I,I 118 | NP_001269241.1 | |
NM_021806.3 | 1069 | Silent Mutation | ATC,ATT | I,I 118 | NP_068578.2 | |
XM_005274714.3 | 1069 | Silent Mutation | ATC,ATT | I,I 139 | XP_005274771.1 | |
XM_005274716.3 | 1069 | Silent Mutation | ATC,ATT | I,I 132 | XP_005274773.1 | |
XM_005277879.4 | 1069 | Intron | XP_005277936.1 | |||
XM_006724831.2 | 1069 | Silent Mutation | ATC,ATT | I,I 140 | XP_006724894.1 | |
XM_006724832.3 | 1069 | Silent Mutation | ATC,ATT | I,I 126 | XP_006724895.1 | |
XM_006724833.2 | 1069 | Intron | XP_006724896.1 | |||
XM_006724834.2 | 1069 | Silent Mutation | ATC,ATT | I,I 85 | XP_006724897.1 | |
XM_011531185.2 | 1069 | Silent Mutation | ATC,ATT | I,I 85 | XP_011529487.1 | |
XM_017029699.1 | 1069 | Silent Mutation | ATC,ATT | I,I 85 | XP_016885188.1 | |
XM_017029700.1 | 1069 | Silent Mutation | ATC,ATT | I,I 77 | XP_016885189.1 | |
XM_017029701.1 | 1069 | Silent Mutation | ATC,ATT | I,I 77 | XP_016885190.1 | |
XM_017029702.1 | 1069 | Silent Mutation | ATC,ATT | I,I 77 | XP_016885191.1 |