Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTTATTTGGACATTGCCAAAGG[C/T]GCTGTGGAGTTATTCTTGAAGGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
INTS6L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INTS6L - integrator complex subunit 6 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182540.4 | 453 | Silent Mutation | GGC,GGT | G,G 30 | NP_872346.3 | |
XM_005262394.2 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_005262451.1 | |
XM_006724740.2 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_006724803.1 | |
XM_006724741.3 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_006724804.1 | |
XM_011531311.2 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_011529613.1 | |
XM_017029342.1 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_016884831.1 | |
XM_017029343.1 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_016884832.1 | |
XM_017029344.1 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_016884833.1 | |
XM_017029345.1 | 453 | UTR 5 | XP_016884834.1 | |||
XM_017029346.1 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_016884835.1 | |
XM_017029347.1 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_016884836.1 | |
XM_017029348.1 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_016884837.1 | |
XM_017029349.1 | 453 | Silent Mutation | GGC,GGT | G,G 30 | XP_016884838.1 |
INTS6L-AS1 - INTS6L antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |