Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCACCATTGATTTACTTTTTGC[C/T]GATGTCTCCATGAAAGGACAGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAP2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAP2C - RAP2C, member of RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271186.1 | 1362 | Silent Mutation | TCA,TCG | S,S 146 | NP_001258115.1 | |
NM_001271187.1 | 1362 | Silent Mutation | TCA,TCG | S,S 80 | NP_001258116.1 | |
NM_021183.4 | 1362 | Silent Mutation | TCA,TCG | S,S 146 | NP_067006.3 | |
XM_006724775.2 | 1362 | Silent Mutation | TCA,TCG | S,S 80 | XP_006724838.1 | |
XM_011531376.1 | 1362 | Silent Mutation | TCA,TCG | S,S 146 | XP_011529678.1 |
RAP2C-AS1 - RAP2C antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |