Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCATCCTTTCCTTACAGTTATC[A/G]CTACAGACTCGTGAGTGCCAGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP1B4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP1B4 - ATPase Na+/K+ transporting family member beta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142447.2 | 137 | Missense Mutation | CAC,CGC | H,R 18 | NP_001135919.1 | |
NM_012069.4 | 137 | Missense Mutation | CAC,CGC | H,R 18 | NP_036201.1 | |
XM_017029381.1 | 137 | Missense Mutation | CAC,CGC | H,R 18 | XP_016884870.1 |