Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTGATGATTCAGAAGCTCCCGA[C/T]GACAACAGTGATGATTCGGAAGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300369 | ||||||||||||||||||||
Literature Links: |
ACRC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACRC - acidic repeat containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052957.4 | 771 | Silent Mutation | GAC,GAT | D,D 150 | NP_443189.1 | |
XM_006724716.3 | 771 | Silent Mutation | GAC,GAT | D,D 150 | XP_006724779.1 | |
XM_011531063.2 | 771 | Silent Mutation | GAC,GAT | D,D 159 | XP_011529365.1 | |
XM_011531064.1 | 771 | Silent Mutation | GAC,GAT | D,D 150 | XP_011529366.1 | |
XM_011531066.2 | 771 | Silent Mutation | GAC,GAT | D,D 132 | XP_011529368.1 | |
XM_011531067.2 | 771 | Silent Mutation | GAC,GAT | D,D 132 | XP_011529369.1 | |
XM_017029965.1 | 771 | Intron | XP_016885454.1 |