Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATATTTTTGTTCTCTCTTCTA[C/T]CAGGTTGTTTGCTATTATTTTCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MORF4L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MORF4L2 - mortality factor 4 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142418.1 | Intron | NP_001135890.1 | ||||
NM_001142419.1 | Intron | NP_001135891.1 | ||||
NM_001142420.1 | Intron | NP_001135892.1 | ||||
NM_001142421.1 | Intron | NP_001135893.1 | ||||
NM_001142422.1 | Intron | NP_001135894.1 | ||||
NM_001142423.1 | Intron | NP_001135895.1 | ||||
NM_001142424.1 | Intron | NP_001135896.1 | ||||
NM_001142425.1 | Intron | NP_001135897.1 | ||||
NM_001142426.1 | Intron | NP_001135898.1 | ||||
NM_001142427.1 | Intron | NP_001135899.1 | ||||
NM_001142428.1 | Intron | NP_001135900.1 | ||||
NM_001142429.1 | Intron | NP_001135901.1 | ||||
NM_001142430.1 | Intron | NP_001135902.1 | ||||
NM_001142431.1 | Intron | NP_001135903.1 | ||||
NM_001142432.1 | Intron | NP_001135904.1 | ||||
NM_012286.2 | Intron | NP_036418.1 |
MORF4L2-AS1 - MORF4L2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |