Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTCTTCCAGGGATGCCTGCTC[C/T]TCCTGAAGAGATGGTTCTCGAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300420 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PJA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PJA1 - praja ring finger ubiquitin ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032396.2 | 1168 | Silent Mutation | GAA,GAG | E,E 396 | NP_001027568.1 | |
NM_022368.4 | 1168 | Silent Mutation | GAA,GAG | E,E 263 | NP_071763.2 | |
NM_145119.3 | 1168 | Silent Mutation | GAA,GAG | E,E 451 | NP_660095.1 | |
XM_005262292.1 | 1168 | Silent Mutation | GAA,GAG | E,E 396 | XP_005262349.1 | |
XM_011531011.2 | 1168 | Silent Mutation | GAA,GAG | E,E 451 | XP_011529313.1 |