Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCAGCCGATTCACAGCATGCATT[A/G]ATCTGGTATTCATGGATGACAACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ADGRG4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRG4 - adhesion G protein-coupled receptor G4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153834.3 | 207 | Missense Mutation | AAT,GAT | N,D 60 | NP_722576.3 | |
XM_011531269.2 | 207 | Missense Mutation | AAT,GAT | N,D 60 | XP_011529571.1 | |
XM_011531271.2 | 207 | UTR 5 | XP_011529573.1 |