Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCTGCTGAGGTCCTGAGATGGA[A/G]GCAAAGACTTCGTTGTCCCAAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXorf23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXorf23 - chromosome X open reading frame 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198279.3 | 5430 | UTR 3 | NP_938020.2 | |||
XM_005274473.3 | 5430 | UTR 3 | XP_005274530.1 | |||
XM_005274474.3 | 5430 | UTR 3 | XP_005274531.1 | |||
XM_005274475.3 | 5430 | UTR 3 | XP_005274532.1 | |||
XM_011545475.2 | 5430 | UTR 3 | XP_011543777.1 | |||
XM_011545476.2 | 5430 | UTR 3 | XP_011543778.1 | |||
XM_011545477.2 | 5430 | Intron | XP_011543779.1 | |||
XM_011545478.2 | 5430 | UTR 3 | XP_011543780.1 | |||
XM_011545479.2 | 5430 | UTR 3 | XP_011543781.1 | |||
XM_011545482.2 | 5430 | UTR 3 | XP_011543784.1 | |||
XM_017029390.1 | 5430 | UTR 3 | XP_016884879.1 | |||
XM_017029391.1 | 5430 | UTR 3 | XP_016884880.1 | |||
XM_017029392.1 | 5430 | Intron | XP_016884881.1 |