Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCTTCATCTAAGAGAAGGAAGA[A/G]TATGTCCTCGTGGTTAGACAAACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300359 | ||||||||||||||||||||
Literature Links: |
CT45A10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CT45A10 - cancer/testis antigen family 45, member A10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAGE1 - sarcoma antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018666.2 | 366 | Missense Mutation | AAT,AGT | N,S 64 | NP_061136.2 | |
XM_017029621.1 | 366 | Missense Mutation | AAT,AGT | N,S 64 | XP_016885110.1 | |
XM_017029622.1 | 366 | Intron | XP_016885111.1 | |||
XM_017029623.1 | 366 | Missense Mutation | AAT,AGT | N,S 64 | XP_016885112.1 | |
XM_017029624.1 | 366 | Missense Mutation | AAT,AGT | N,S 64 | XP_016885113.1 | |
XM_017029625.1 | 366 | Missense Mutation | AAT,AGT | N,S 64 | XP_016885114.1 | |
XM_017029626.1 | 366 | Intron | XP_016885115.1 | |||
XM_017029627.1 | 366 | Intron | XP_016885116.1 |