Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATTTGAAAGTTGGTGAGGGCAT[G/T]CAGTTTTGGTAGGAATGTGAATCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300883 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNX12 - sorting nexin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256185.1 | Intron | NP_001243114.1 | ||||
NM_001256186.1 | Intron | NP_001243115.1 | ||||
NM_001256187.1 | Intron | NP_001243116.1 | ||||
NM_001256188.1 | Intron | NP_001243117.1 | ||||
NM_013346.3 | Intron | NP_037478.2 |