Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATCTCCCTTTCATTCCGATGTG[A/G]TCATGTAAACTCCATAAACTTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300838 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMPD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMPD4 - FERM and PDZ domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014728.3 | Intron | NP_055543.2 | ||||
XM_005274632.3 | Intron | XP_005274689.2 | ||||
XM_011545613.2 | Intron | XP_011543915.1 | ||||
XM_017029983.1 | Intron | XP_016885472.1 | ||||
XM_017029984.1 | Intron | XP_016885473.1 | ||||
XM_017029985.1 | Intron | XP_016885474.1 | ||||
XM_017029986.1 | Intron | XP_016885475.1 |