Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTATGCATCAAATGAAGAGAGC[C/T]GAAGTCAGTGGTTGAAAGCATTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300335 MIM: 300101 | ||||||||||||||||||||
Literature Links: |
ACE2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACE2 - angiotensin I converting enzyme 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021804.2 | 468 | Intron | NP_068576.1 | |||
XM_011545549.1 | 468 | Intron | XP_011543851.1 | |||
XM_011545551.2 | 468 | Intron | XP_011543853.1 | |||
XM_011545552.1 | 468 | Intron | XP_011543854.1 |
BMX - BMX non-receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320866.1 | 468 | Nonsense Mutation | CGA,TGA | R,* 99 | NP_001307795.1 | |
NM_001721.6 | 468 | Nonsense Mutation | CGA,TGA | R,* 99 | NP_001712.1 | |
NM_203281.2 | 468 | Nonsense Mutation | CGA,TGA | R,* 99 | NP_975010.1 | |
XM_017029752.1 | 468 | Nonsense Mutation | CGA,TGA | R,* 99 | XP_016885241.1 |