Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCAATGGTCAAGCTCCATAGCA[G/T]AAGAATTGGTATTCCTCATGATGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300566 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHFPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LHFPL1 - lipoma HMGIC fusion partner-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178175.3 | 1172 | Missense Mutation | TAT,TCT | Y,S 210 | NP_835469.1 | |
XM_011530943.2 | 1172 | Missense Mutation | TAT,TCT | Y,S 233 | XP_011529245.1 | |
XM_011530944.2 | 1172 | Missense Mutation | TAT,TCT | Y,S 210 | XP_011529246.1 | |
XM_011530946.2 | 1172 | Missense Mutation | TAT,TCT | Y,S 177 | XP_011529248.1 | |
XM_017029485.1 | 1172 | Missense Mutation | TAT,TCT | Y,S 233 | XP_016884974.1 |