Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATACTGGGTCCAGACCTGACAG[A/G]AGGGAAGAGACCAGCATTGGGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMCX5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMCX5 - armadillo repeat containing, X-linked 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168478.1 | 1114 | Silent Mutation | AGA,AGG | R,R 152 | NP_001161950.1 | |
NM_001168479.1 | 1114 | Silent Mutation | AGA,AGG | R,R 152 | NP_001161951.1 | |
NM_001168480.1 | 1114 | Silent Mutation | AGA,AGG | R,R 152 | NP_001161952.1 | |
NM_001168482.1 | 1114 | Silent Mutation | AGA,AGG | R,R 152 | NP_001161954.1 | |
NM_001168485.1 | 1114 | Silent Mutation | AGA,AGG | R,R 152 | NP_001161957.1 | |
NM_022838.3 | 1114 | Silent Mutation | AGA,AGG | R,R 152 | NP_073749.2 |
ARMCX5-GPRASP2 - ARMCX5-GPRASP2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199818.1 | 1114 | Intron | NP_001186747.1 |