Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCACTGTGAACCACTAGGTGGGG[C/T]GTACAGGATGCAGCAGTTGTCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300012 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMARCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMARCA1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282874.1 | Intron | NP_001269803.1 | ||||
NM_001282875.1 | Intron | NP_001269804.1 | ||||
NM_003069.4 | Intron | NP_003060.2 | ||||
XM_005262461.2 | Intron | XP_005262518.1 | ||||
XM_005262462.2 | Intron | XP_005262519.1 | ||||
XM_006724782.2 | Intron | XP_006724845.1 | ||||
XM_017029750.1 | Intron | XP_016885239.1 | ||||
XM_017029751.1 | Intron | XP_016885240.1 |