Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCGGGTTAGACAACGATGTTGG[C/G]GTGGAGGTAGGCGGCAGTGGCGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300961 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MID1IP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MID1IP1 - MID1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098790.1 | 2358 | Silent Mutation | GGC,GGG | G,G 53 | NP_001092260.1 | |
NM_001098791.1 | 2358 | Silent Mutation | GGC,GGG | G,G 53 | NP_001092261.1 | |
NM_021242.5 | 2358 | Silent Mutation | GGC,GGG | G,G 53 | NP_067065.1 |
MID1IP1-AS1 - MID1IP1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |