Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAGGTATCTTAGATCTTCCTTAG[G/T]GATCTCATTAAGAATATCCGAAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300820 MIM: 300502 | ||||||||||||||||||||
Literature Links: |
MAP3K15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K15 - mitogen-activated protein kinase kinase kinase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001671.3 | 3149 | Missense Mutation | ACT,CCT | T,P 1276 | NP_001001671.3 | |
XM_011545507.2 | 3149 | Missense Mutation | ACT,CCT | T,P 1287 | XP_011543809.2 | |
XM_011545508.2 | 3149 | Missense Mutation | ACT,CCT | T,P 1258 | XP_011543810.2 | |
XM_011545510.2 | 3149 | Missense Mutation | ACT,CCT | T,P 834 | XP_011543812.1 | |
XM_011545511.2 | 3149 | Missense Mutation | ACT,CCT | T,P 711 | XP_011543813.1 | |
XM_017029511.1 | 3149 | Intron | XP_016885000.1 |
PDHA1 - pyruvate dehydrogenase (lipoamide) alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000284.3 | 3149 | UTR 3 | NP_000275.1 | |||
NM_001173454.1 | 3149 | UTR 3 | NP_001166925.1 | |||
NM_001173455.1 | 3149 | UTR 3 | NP_001166926.1 | |||
NM_001173456.1 | 3149 | UTR 3 | NP_001166927.1 | |||
XM_017029574.1 | 3149 | Intron | XP_016885063.1 |