Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGGCCTCCAACGAAGATTTCTC[C/T]ATCACACAAGACCTGGAGATCCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300294 MIM: 300570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBTPS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBTPS2 - membrane bound transcription factor peptidase, site 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015884.3 | 522 | Intron | NP_056968.1 |
YY2 - YY2 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206923.3 | 522 | Silent Mutation | TCC,TCT | S,S 8 | NP_996806.2 |