Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAACCACTGGTCCTTTTGTTGC[C/T]GGATGAAGCTGGTATAGTGGCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP51 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP51 - ubiquitin specific peptidase 51 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_201286.3 | 2091 | Missense Mutation | CAG,CGG | Q,R 671 | NP_958443.1 | |
XM_017029299.1 | 2091 | Missense Mutation | CAG,CGG | Q,R 671 | XP_016884788.1 | |
XM_017029300.1 | 2091 | Missense Mutation | CAG,CGG | Q,R 671 | XP_016884789.1 | |
XM_017029301.1 | 2091 | Missense Mutation | CAG,CGG | Q,R 671 | XP_016884790.1 |