Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATATGGCAAAGAGCACAGGAAA[A/G]ATGATGCGGGAAATTTTGTCAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 305660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRA3 - gamma-aminobutyric acid type A receptor alpha3 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000808.3 | 1414 | Silent Mutation | ATC,ATT | I,I 462 | NP_000799.1 | |
XM_006724811.2 | 1414 | UTR 3 | XP_006724874.1 |