Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGGGCTCCGACTGCTCTGATGC[A/G]GAATCCGAGGCTGACCGCGACTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300134 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP9 - dual specificity phosphatase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318503.1 | 639 | Silent Mutation | GCA,GCG | A,A 174 | NP_001305432.1 | |
NM_001395.3 | 639 | Silent Mutation | GCA,GCG | A,A 174 | NP_001386.1 | |
XM_011531123.1 | 639 | Silent Mutation | GCA,GCG | A,A 205 | XP_011529425.1 | |
XM_011531124.1 | 639 | Silent Mutation | GCA,GCG | A,A 174 | XP_011529426.1 |