Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCGCCACCAAGACCGTCATCTA[C/T]CTGCTCAACCCCTGTCACCTGGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM164 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM164 - transmembrane protein 164 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017698.2 | 988 | Intron | NP_060168.2 | |||
NM_032227.3 | 988 | Silent Mutation | TAC,TAT | Y,Y 118 | NP_115603.2 | |
XM_005262205.4 | 988 | Silent Mutation | TAC,TAT | Y,Y 118 | XP_005262262.1 | |
XM_005262206.3 | 988 | Silent Mutation | TAC,TAT | Y,Y 118 | XP_005262263.1 | |
XM_005262208.4 | 988 | UTR 5 | XP_005262265.1 | |||
XM_011531051.2 | 988 | UTR 5 | XP_011529353.1 | |||
XM_011531054.1 | 988 | UTR 5 | XP_011529356.1 | |||
XM_011531055.2 | 988 | Intron | XP_011529357.1 | |||
XM_017029894.1 | 988 | Intron | XP_016885383.1 | |||
XM_017029895.1 | 988 | Silent Mutation | TAC,TAT | Y,Y 118 | XP_016885384.1 | |
XM_017029896.1 | 988 | UTR 5 | XP_016885385.1 | |||
XM_017029897.1 | 988 | Intron | XP_016885386.1 | |||
XM_017029898.1 | 988 | Silent Mutation | TAC,TAT | Y,Y 118 | XP_016885387.1 | |
XM_017029899.1 | 988 | UTR 5 | XP_016885388.1 | |||
XM_017029900.1 | 988 | UTR 5 | XP_016885389.1 |