Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176705 | ||||||||||||||||||||
Literature Links: |
PHB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHB - prohibitin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281496.1 | 496 | Intron | NP_001268425.1 | |||
NM_001281497.1 | 496 | Intron | NP_001268426.1 | |||
NM_001281715.1 | 496 | Intron | NP_001268644.1 | |||
NM_002634.3 | 496 | Intron | NP_002625.1 | |||
XM_017024762.1 | 496 | Silent Mutation | CAA,CAG | Q,Q 142 | XP_016880251.1 | |
XM_017024763.1 | 496 | Intron | XP_016880252.1 |