Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607521 MIM: 104750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPS5 - HPS5, biogenesis of lysosomal organelles complex 2 subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAA1 - serum amyloid A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000331.5 | 460 | Missense Mutation | AAA,AGA | K,R 108 | NP_000322.2 | |
NM_001178006.2 | 460 | Missense Mutation | AAA,AGA | K,R 108 | NP_001171477.1 | |
NM_199161.4 | 460 | Missense Mutation | AAA,AGA | K,R 108 | NP_954630.1 |
ST13P5 - suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |