Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611596 MIM: 616925 | ||||||||||||||||||||
Literature Links: |
DHRS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHRS4 - dehydrogenase/reductase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282987.1 | 62 | Missense Mutation | AAG,ATG | K,M 3 | NP_001269916.1 | |
NM_001282988.1 | 62 | Missense Mutation | AAG,ATG | K,M 3 | NP_001269917.1 | |
NM_001282989.1 | 62 | Missense Mutation | AAG,ATG | K,M 3 | NP_001269918.1 | |
NM_001282990.1 | 62 | Missense Mutation | AAG,ATG | K,M 3 | NP_001269919.1 | |
NM_001282991.1 | 62 | Missense Mutation | AAG,ATG | K,M 3 | NP_001269920.1 | |
NM_021004.3 | 62 | Missense Mutation | AAG,ATG | K,M 3 | NP_066284.2 | |
XM_006720005.3 | 62 | Missense Mutation | AAG,ATG | K,M 3 | XP_006720068.1 | |
XM_011536366.2 | 62 | Missense Mutation | AAG,ATG | K,M 3 | XP_011534668.1 |
DHRS4-AS1 - DHRS4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |