Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATCATTGAAGCTGCTCTCAGGG[T/C]TCCCCTTGGCCTGAGCAGGGCCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 124030 | ||||||||||||||||||||
Literature Links: |
CYP2D6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP2D6 - cytochrome P450 family 2 subfamily D member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000106.5 | 791 | Missense Mutation | AAC,AGC | N,S 285 | NP_000097.3 | |
NM_001025161.2 | 791 | Missense Mutation | AAC,AGC | N,S 234 | NP_001020332.2 | |
XM_011529966.2 | 791 | Intron | XP_011528268.1 | |||
XM_011529968.2 | 791 | Intron | XP_011528270.1 | |||
XM_011529970.2 | 791 | Intron | XP_011528272.1 | |||
XM_011529972.2 | 791 | Intron | XP_011528274.1 |
LOC102723722 - uncharacterized LOC102723722 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017029174.1 | 791 | Intron | XP_016884663.1 |
NDUFA6-AS1 - NDUFA6 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |