Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600169 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MICA - MHC class I polypeptide-related sequence A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000247.2 | 652 | Missense Mutation | AAA,GAA | K,E 196 | NP_000238.1 | |
NM_001177519.2 | 652 | Missense Mutation | AAA,GAA | K,E 196 | NP_001170990.1 | |
NM_001289152.1 | 652 | Missense Mutation | AAA,GAA | K,E 99 | NP_001276081.1 | |
NM_001289153.1 | 652 | Missense Mutation | AAA,GAA | K,E 99 | NP_001276082.1 | |
NM_001289154.1 | 652 | Missense Mutation | AAA,GAA | K,E 58 | NP_001276083.1 |
Set Membership: |
HapMap |