Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTGGTGAAGTTGGTTCGGGAGC[T/G]GTATGTGTGCTCAGGCACCCCCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 188230 | ||||||||||||||||||||
Literature Links: |
THY1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THY1 - Thy-1 cell surface antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311160.1 | 1263 | Missense Mutation | AGC,CGC | S,R 75 | NP_001298089.1 | |
NM_001311162.1 | 1263 | Missense Mutation | AGC,CGC | S,R 75 | NP_001298091.1 | |
NM_006288.4 | 1263 | Missense Mutation | AGC,CGC | S,R 75 | NP_006279.2 |
USP2-AS1 - USP2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |