Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCATTCACCAGTGCTGGGGTCTT[T/C]TCCACAAGGTTACGGACAAATGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613213 | ||||||||||||||||||||
Literature Links: |
ATP5L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP5L2 - ATP synthase, H+ transporting, mitochondrial Fo complex subunit G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165877.1 | 357 | Silent Mutation | GAA,GAG | E,E 10 | NP_001159349.1 |
CYB5R3 - cytochrome b5 reductase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000398.6 | 357 | Intron | NP_000389.1 | |||
NM_001129819.2 | 357 | Intron | NP_001123291.1 | |||
NM_001171660.1 | 357 | Intron | NP_001165131.1 | |||
NM_001171661.1 | 357 | Intron | NP_001165132.1 | |||
NM_007326.4 | 357 | Intron | NP_015565.1 |