Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGGGAATCCAGTATACCCTCA[C/G]ACCTTGCATCTTCCTCAGGCTCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607431 | ||||||||||||||||||||
Literature Links: |
DAZAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DAZAP2 - DAZ associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136264.1 | 259 | Silent Mutation | CAC,CAG | H,Q 25 | NP_001129736.1 | |
NM_001136266.1 | 259 | Silent Mutation | CAC,CAG | H,Q 25 | NP_001129738.1 | |
NM_001136267.1 | 259 | Silent Mutation | CAC,CAG | H,Q 25 | NP_001129739.1 | |
NM_001136268.1 | 259 | Silent Mutation | CAC,CAG | H,Q 25 | NP_001129740.1 | |
NM_001136269.1 | 259 | Silent Mutation | CAC,CAG | H,Q 25 | NP_001129741.1 | |
NM_014764.3 | 259 | Silent Mutation | CAC,CAG | H,Q 25 | NP_055579.1 |
SMAGP - small cell adhesion glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |