Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603703 | ||||||||||||||||||||
Literature Links: |
RPL6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPL6 - ribosomal protein L6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000970.4 | 890 | Intron | NP_000961.2 | |||
NM_001024662.2 | 890 | Intron | NP_001019833.1 | |||
NM_001320137.1 | 890 | Intron | NP_001307066.1 | |||
NM_001320138.1 | 890 | Missense Mutation | AGC,ATC | S,I 204 | NP_001307067.1 | |
NM_001320139.1 | 890 | Intron | NP_001307068.1 | |||
NM_001320140.1 | 890 | Missense Mutation | AGC,ATC | S,I 204 | NP_001307069.1 | |
NM_001320141.1 | 890 | Missense Mutation | AGC,ATC | S,I 204 | NP_001307070.1 | |
NM_001320142.1 | 890 | Intron | NP_001307071.1 | |||
XM_017019781.1 | 890 | Missense Mutation | AGC,ATC | S,I 204 | XP_016875270.1 | |
XM_017019782.1 | 890 | Intron | XP_016875271.1 |