Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603637 MIM: 611333 MIM: 140750 MIM: 612000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL27A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPL27A - ribosomal protein L27a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000990.4 | 651 | Silent Mutation | ACG,ACT | T,T 95 | NP_000981.1 |
SNORA3A - small nucleolar RNA, H/ACA box 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA3B - small nucleolar RNA, H/ACA box 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ST5 - suppression of tumorigenicity 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM66 - tripartite motif containing 66 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |