Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVPLL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EVPLL - envoplakin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145127.1 | 1139 | Silent Mutation | CCA,CCG | P,P 296 | NP_001138599.1 | |
XM_011523982.2 | 1139 | Silent Mutation | CCA,CCG | P,P 293 | XP_011522284.1 | |
XM_011523983.2 | 1139 | Intron | XP_011522285.1 | |||
XM_011523985.2 | 1139 | Silent Mutation | CCA,CCG | P,P 264 | XP_011522287.1 | |
XM_011523986.2 | 1139 | Intron | XP_011522288.1 | |||
XM_011523987.2 | 1139 | Intron | XP_011522289.1 |
Set Membership: |
HapMap |