Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176397 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSG8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSG8 - pregnancy specific beta-1-glycoprotein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130167.1 | 527 | Silent Mutation | TAC,TAT | Y,Y 143 | NP_001123639.1 | |
NM_001130168.1 | 527 | Intron | NP_001123640.1 | |||
NM_182707.2 | 527 | Silent Mutation | TAC,TAT | Y,Y 143 | NP_874366.1 | |
XM_011526986.1 | 527 | Intron | XP_011525288.1 | |||
XM_011526987.2 | 527 | Intron | XP_011525289.1 | |||
XM_011526988.2 | 527 | Intron | XP_011525290.1 |