Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605093 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SH2B3 - SH2B adaptor protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291424.1 | 540 | Intron | NP_001278353.1 | |||
NM_005475.2 | 540 | Missense Mutation | ACC,AGC | T,S 165 | NP_005466.1 | |
XM_005253818.4 | 540 | Missense Mutation | ACC,AGC | T,S 165 | XP_005253875.1 | |
XM_005253819.4 | 540 | Missense Mutation | ACC,AGC | T,S 165 | XP_005253876.1 | |
XM_006719180.3 | 540 | Intron | XP_006719243.1 | |||
XM_011537719.2 | 540 | Missense Mutation | ACC,AGC | T,S 165 | XP_011536021.1 | |
XM_011537720.2 | 540 | Missense Mutation | ACC,AGC | T,S 165 | XP_011536022.1 | |
XM_011537721.2 | 540 | Intron | XP_011536023.1 |