Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616862 | ||||||||||||||||||||
Literature Links: |
RPL34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPL34 - ribosomal protein L34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000995.4 | 385 | Silent Mutation | GCA,GCC | A,A 111 | NP_000986.2 | |
NM_001319232.1 | 385 | Intron | NP_001306161.1 | |||
NM_001319234.1 | 385 | Silent Mutation | GCA,GCC | A,A 111 | NP_001306163.1 | |
NM_001319235.1 | 385 | Intron | NP_001306164.1 | |||
NM_001319236.1 | 385 | Intron | NP_001306165.1 | |||
NM_033625.3 | 385 | Silent Mutation | GCA,GCC | A,A 111 | NP_296374.1 |
RPL34-AS1 - RPL34 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |