Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600182 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6775 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6775 - microRNA 6775 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A5 - solute carrier family 7 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006721286.3 | 1677 | UTR 3 | XP_006721349.2 | |||
XM_017023735.1 | 1677 | Intron | XP_016879224.1 | |||
XM_017023736.1 | 1677 | Intron | XP_016879225.1 | |||
XM_017023737.1 | 1677 | Intron | XP_016879226.1 |
Set Membership: |
HapMap |