Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTGTACAGACATAGATGCTCAC[C/T]GTTCAGTTGGTTGAAGACTGGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600874 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GNG5 - G protein subunit gamma 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA1 - spermatogenesis associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310156.1 | Intron | NP_001297085.1 | ||||
XM_011542514.2 | Intron | XP_011540816.1 | ||||
XM_011542515.2 | Intron | XP_011540817.1 | ||||
XM_011542516.1 | Intron | XP_011540818.1 | ||||
XM_011542517.1 | Intron | XP_011540819.1 | ||||
XM_011542519.2 | Intron | XP_011540821.1 | ||||
XM_011542520.2 | Intron | XP_011540822.1 | ||||
XM_011542521.2 | Intron | XP_011540823.1 | ||||
XM_011542522.2 | Intron | XP_011540824.1 |