Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGCCATTGCAGGTCTCAAGCC[A/G]TTAGGTTTCATTTTCCTCCTCCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604650 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFIT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFIT3 - interferon induced protein with tetratricopeptide repeats 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031683.3 | 17 | Intron | NP_001026853.1 | |||
NM_001289758.1 | 17 | UTR 5 | NP_001276687.1 | |||
NM_001289759.1 | 17 | Intron | NP_001276688.1 | |||
NM_001549.5 | 17 | UTR 5 | NP_001540.2 |
Set Membership: |
HapMap |