Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAGAAGAGCGGGCTGTGATACC[C/T]CCTCGACGCCGTCAAACACTGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604825 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FEZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FEZ1 - fasciculation and elongation protein zeta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005103.4 | 176 | Intron | NP_005094.1 | |||
NM_022549.3 | 176 | Intron | NP_072043.1 | |||
XM_005271734.2 | 176 | UTR 5 | XP_005271791.1 | |||
XM_005271735.2 | 176 | Intron | XP_005271792.1 |
LOC403312 - putative uncharacterized protein MGC39545 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301851.1 | 176 | Missense Mutation | CCC,TCC | P,S 21 | NP_001288780.1 |