Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTAGTCTCCTTTAAAACAGACAC[A/G]AAGGAGGCAGAGAGAAAAAATTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609673 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDGFD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDGFD - platelet derived growth factor D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025208.4 | 2864 | UTR 3 | NP_079484.1 | |||
NM_033135.3 | 2864 | UTR 3 | NP_149126.1 |
Set Membership: |
HapMap |