Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTGAGGTTTTCTTAATGGCCTA[A/G]TGTATGGCCTTGAAGAGTGTTCCGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615126 MIM: 606780 | |||||||||||||||||||||||
Literature Links: |
ANKRD13D PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ANKRD13D - ankyrin repeat domain 13D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207354.2 | Intron | NP_997237.2 | ||||
XM_011544985.1 | Intron | XP_011543287.1 | ||||
XM_011544986.1 | Intron | XP_011543288.1 | ||||
XM_011544987.2 | Intron | XP_011543289.1 | ||||
XM_011544988.2 | Intron | XP_011543290.1 |
SSH3 - slingshot protein phosphatase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |