Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179060 MIM: 611450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDHB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDHB - pyruvate dehydrogenase (lipoamide) beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000925.3 | 1425 | UTR 3 | NP_000916.2 | |||
NM_001173468.1 | 1425 | UTR 3 | NP_001166939.1 | |||
NM_001315536.1 | 1425 | UTR 3 | NP_001302465.1 |
PXK - PX domain containing serine/threonine kinase like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |